Learn More
Sino Biological Anti-PTPN9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11517T20100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PTPN9 / PTP-MEG2 (rh PTPN9 / PTP-MEG2; Catalog#11517-H20B; NP_002824.1; Ala285-Gln593). PTPN9 / PTP-MEG2 specific IgG was purified by Human PTPN9 / PTP-MEG2 affinity chromatography.
Catalog No. 50-255-4703