Learn More
Sino Biological Anti-CD45 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100342T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CD45 / PTPRC, and purified by antigen affinity chromatography.
Catalog No. 50-241-2250