Learn More
Sino Biological Recombinant Human CD45 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 14197H49HB20
Encompass_Preferred
A DNA sequence encoding the Human PTPRC (NP_002829.3) (Gln26-Lys577) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4144