Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PTPRD Synthetic Peptide

Catalog No. PEP1435
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1435 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1435 Supplier Invitrogen™ Supplier No. PEP1435
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 18 amino acid peptide near the amino terminus of human PTPRD. PEP-1435 can be used as a blocking peptide with polyclonal antibody PA5-34392.

PTPRD is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular region, a single transmembrane segment and two tandem intracytoplasmic catalytic domains, and thus represents a receptor-type PTP. The extracellular region of this protein is composed of three Ig-like and eight fibronectin type III-like domains. Studies of the similar genes in chicken and fly suggest the role of this PTP is in promoting neurite growth, and regulating neurons axon guidance.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 5789
Species Human
Purification Method purified
Accession Number P23468
Common Name PTPRD
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 1110002J03Rik; 3000002J10Rik; B230219D21Rik; HPTP; HPTPD; HPTPDELTA; protein tyrosine phosphatase, receptor type D; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, delta polypeptide; Protein-tyrosine phosphatase delta; PTPD; Ptprd; receptor-type tyrosine-protein phosphatase delta; RPTPDELTA; R-PTP-delta
Gene Symbol PTPRD
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.