Learn More
Sino Biological Anti-DEP-1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13165MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DEP1 / PTPRJ (rh DEP1 / PTPRJ; Catalog#13165-H07E; Q12913; Arg997-Ala1337). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4297