Learn More
Sino Biological Anti-DEP-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13165T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DEP1 / PTPRJ (rh DEP1 / PTPRJ; Catalog#13165-H07E; Q12913; Arg997-Ala1337). DEP1 / PTPRJ specific IgG was purified by Human DEP1 / PTPRJ affinity chromatography.
Catalog No. 50-255-4296