Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PTX-3 ELISA Kit

Catalog No. EH386RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH386RB 96 Tests
EH386RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH386RB Supplier Invitrogen™ Supplier No. EH386RB
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human PTX-3 quantitates human PTX-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PTX-3.

Pentraxins are a protein superfamily that is characterized by a cyclic multimeric structure. Ptx3, also known as tumor necrosis factor-stimulated gene sequence-14 (TSG14), is a secreted pattern-recognition receptor that has a non-redundant role in resistance to selected microbial agents. Ptx3 belongs to the family of 'long pentraxins', which have C-terminal pentraxin domains and novel amino-terminal domains. Ptx3 binds selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa and Salmonella typhimurium. It is synthesized in IgA glomerulonephritis and activates mesangial cells. Secretion of Ptx3 in adipose cells can be induced by TNF. Ptx3 is also involved in amplification of inflammatory reactions and regulation of innate immunity. The human PTX3 gene maps to chromosome 3q25.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P26022
Assay Range 0.08-20 ng/mL
Assay Sensitivity 0.08 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI607804; long pentraxin 3; pentaxin related; pentaxin-related protein PTX3; pentraxin 3; pentraxin 3, long; pentraxin related; pentraxin related gene; pentraxin-related protein PTX3; Ptx3; TNF alpha-induced protein 5; TNFAIP5; Tsg14; TSG-14; Tumor necrosis factor alpha-induced protein 5; tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14AI607804; long pentraxin 3; pentaxin related; pentaxin-related protein PTX3; pentraxin 3; pentraxin 3, long; pentraxin related; pentraxin related gene; pentraxin-related protein PTX3; Ptx3; TNF alpha-induced protein 5; TNFAIP5; Tsg14; TSG-14; Tumor necrosis factor alpha-induced protein 5; tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14;
Gene ID (Entrez) 5806
Gene Symbol PTX3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target PTX3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.