Learn More
Sino Biological Anti-Pentraxin 3/TSG-14 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12082MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PTX3/Pentraxin 3/TSG-14 (rh PTX3/Pentraxin 3/TSG-14; Catalog#12082-H08H; NP_002843.2; Met1-Ser381). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0455