Learn More
Sino Biological Recombinant Anti-Pentraxin 3/TSG-14 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12082R07020
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PTX3 / Pentraxin 3 / TSG-14 (rh PTX3 / Pentraxin 3 / TSG-14; Catalog#12082-H08H; NP_002843.2; Met 1-Ser 381).
Catalog No. 50-255-4464