Learn More
Sino Biological Anti-Pentraxin 3/TSG-14 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12082RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PTX3 / Pentraxin 3 / TSG-14 (rh PTX3 / Pentraxin 3 / TSG-14; Catalog#12082-H08H; NP_002843.2; Met 1-Ser 381). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8764