Learn More
Biomatik Corporation Human Pulmonary Activation Regulated Chemokine (PARC) CLIA Kit
Supplier:  Biomatik Corporation EKU09665
Encompass_Preferred
Human Pulmonary Activation Regulated Chemokine (PARC) CLIA Kit for detecting PARC in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.27-200ng/ml. Sensitivity: 0.13ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P55774. Gene ID: 6362. Target Synonyms: CCL18; AMAC1; SCYA18; DCCK1; MIP-4; CKb7; Alternative Macrophage Activation Associated CC Chemokine 1; Chemokine C-C-Motif Ligand 18; Macrophage inflammatory protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7779