Learn More
Sciencell Research Laboratories Human Pulmonary Alveolar Epithelial Cells, Catalog #3200, >1 x 10^6 cells/vial, 1 ml, Frozen, Cryopreserved, Immunofluorescence for cytokeratin-18/19, Negative for common contaminants
Supplier:  Sciencell Research Laboratories 3200
Encompass
Human Pulmonary Alveolar Epithelial Cells (HPAEpiC) are cryopreserved primary cells sourced from human lung tissue, specifically intended for *in vitro* research applications. These cells represent both Type I and Type II alveolar epithelial cells, integral for lung function and health. HPAEpiC are characterized by specific markers and have undergone rigorous testing for contaminants, ensuring reliability for various pulmonary studies.
  • Primary cell type from human lung tissue
  • Characterized by immunofluorescence for cytokeratin-18 and cytokeratin-19
  • Negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • Recommended for research on pulmonary physiology and injury
  • Not suitable for long-term expansion due to differentiation
  • Optimal growth in Alveolar Epithelial Cell Medium (AEpiCM)
  • Delivery state: Frozen
  • Cells per vial: >1 x 10^6
  • Volume per vial: 1 ml
Catalog No. NC0689020