Learn More
Biomatik Corporation Human Purinergic Receptor P2X, Ligand Gated Ion Channel 7 (P2RX7) CLIA Kit
Supplier:  Biomatik Corporation EKU09782
Encompass_Preferred
Human Purinergic Receptor P2X, Ligand Gated Ion Channel 7 (P2RX7) CLIA Kit for detecting P2RX7 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q99572. Gene ID: 5027. Target Synonyms: P2X7; P2X Purinoceptor 7; ATP receptor; Purinergic receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6442