Learn More
Biomatik Corporation Human Purinergic Receptor P2Y, G Protein Coupled 14 (P2RY14) ELISA Kit
Supplier:  Biomatik Corporation EKU06981
Encompass_Preferred
Human Purinergic Receptor P2Y, G Protein Coupled 14 (P2RY14) ELISA Kit for detecting P2RY14 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15391. Gene ID: 9934. Target Synonyms: GPR105; P2Y14; P2Y Purinoceptor 14; G Protein-Coupled Receptor 105; UDP-glucose receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8791