Learn More
Biomatik Corporation Human Puromycin Sensitive Aminopeptidase (PSA) ELISA Kit
Supplier:  Biomatik Corporation EKU06982
Encompass_Preferred
Human Puromycin Sensitive Aminopeptidase (PSA) ELISA Kit for detecting PSA in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55786-1. Gene ID: 9520. Target Synonyms: NPEPPS; AAP-S; Metalloproteinase MP100; Cytosol alanyl aminopeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7446