Learn More
Sino Biological Anti-CD155/PVR Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10109MM1120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD155 extracellular domain (rh CD155; Catalog#10109-H08H; Met 1-Asn 343; P15151-1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2382