Learn More
CPC Scientific Glp-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH (trifluoroacetate salt) 0.1MG
SDP
Catalog No. 502108665
Encompass
The fibrils in the plaques formed in the brains of patients with Alzheimer?s disease contain significant amounts of truncated Abeta proteins. This fragments contain an N-terminal pyroglutamic acid. The g-carboxy group of glutamic acid forms a cyclic bond with the free N-terminal amino group to form pyroglutamateONE-LETTER SEQUENCE: Glp-FRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVMOLECULAR FORMULA: CHNOSMOLECULAR WEIGHT:4125.68STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [161818-04-8], SYNONYMS: (Pyr)-Amyloid β-Protein (3-40) RESEARCH AREA: Alzheimer's DiseaseREFERENCES: Mok, SS. et al. J. Neurosci. Res. 61, 449 (2000); Russo, C. et al. Nature. 405, 531 (2000)
Catalog No. 50-210-8665 Supplier CPC Scientific Supplier No. AMYD015A
The fibrils in the plaques formed in the brains of patients with Alzheimer?s disease contain significant amounts of truncated Abeta proteins. This fragments contain an N-terminal pyroglutamic acid. The g-carboxy group of glutamic acid forms a cyclic bond with the free N-terminal amino group to form pyroglutamateONE-LETTER SEQUENCE: Glp-FRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVMOLECULAR FORMULA: CHNOSMOLECULAR WEIGHT:4125.68STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [161818-04-8], SYNONYMS: (Pyr)-Amyloid β-Protein (3-40) RESEARCH AREA: Alzheimer's DiseaseREFERENCES: Mok, SS. et al. J. Neurosci. Res. 61, 449 (2000); Russo, C. et al. Nature. 405, 531 (2000)