Learn More
Biomatik Corporation Human Pyruvate Dehydrogenase Phosphatase (PDP) ELISA Kit
Supplier:  Biomatik Corporation EKU06991
Encompass_Preferred
Human Pyruvate Dehydrogenase Phosphatase (PDP) ELISA Kit for detecting PDP in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9P0J1. Gene ID: 54704. Target Synonyms: PDP1; PDH; PPM2C; Protein Phosphatase 2C Magnesium-Dependent Catalytic Subunit; Pyruvate Dehyrogenase Phosphatase Catalytic Subunit 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6478