Learn More
Biomatik Corporation Human Quinoid Dihydropteridine Reductase (QDPR) ELISA Kit
Supplier:  Biomatik Corporation EKU06999
Encompass_Preferred
Human Quinoid Dihydropteridine Reductase (QDPR) ELISA Kit for detecting QDPR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.051ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09417. Gene ID: 5860. Target Synonyms: DHPR; PKU2; SDR33C1; HDHPR; 6,7-Dihydropteridine Reductase; Short Chain Dehydrogenase/Reductase Family 33C,Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6721