Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human R-Spondin-3 Recombinant Protein, PeproTech®

Catalog No. 12044100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12044100UG 100 μg
120-44-1MG 1 mg
120-44-20UG 20 μg
12044250UG 250 μg
12044500UG 500 μg
120-44-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 12044100UG Supplier Gibco™ Supplier No. 12044100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human R-Spondin-3 is a 26.9 kDa protein consisting of 240 amino acid residues. Due to glycosylation, R-Spondin-3 migrates at an apparent molecular weight of approximately 37.0 kDa by SDS PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

RSPO3 is a member of the R-spondin family and plays a role in the regulation of Wnt (wingless-type MMTV integration site family)/beta-catenin and Wnt/planar cell polarity (PCP) signaling pathways, which are involved in development, cell growth and disease pathogenesis. RSPO3 has been shown to be involved in regulating the balance between angioblast and blood cell specification during embryonic vasculogenesis and angiogenesis and may be involved in tumor development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BXY4
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 84870
Molecular Weight (g/mol) 26.9 kDa
Name Human R-Spondin-3
Purification Method Purified
Quantity 100 μg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 2810459H04Rik; AW742308; Cabriolet; cristin 1; Cristin1; cristin-1; cysteine-rich and single thrombospondin domain-containing protein 1; hPWTSR; hRspo3; Nucleopondin; protein with TSP type-1 repeat; PWTSR; RGD1563246; Roof plate-specific spondin-3; Rspo3; R-spondin 3; R-spondin 3 homolog; R-spondin 3 homolog (Xenopus laevis); R-spondin 3-like protein; R-spondin-3; Thrombospondin type-1 domain-containing protein 2; thrombospondin, type I, domain 2; thrombospondin, type I, domain containing 2; Thsd2; Unknown (protein for MGC:137973)
Common Name RSPO3
Gene Symbol RSPO3
Biological Activity R-Spondin-3 enhances BMP-2-mediated differentiation of MC3T3-E1 cells. The expected ED50 for this effect is 0.8 -2.0 ug/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MHPNVSQGCQ GGCATCSDYN GCLSCKPRLF FALERIGMKQ IGVCLSSCPS GYYGTRYPDI NKCTKCKADC DTCFNKNFCT KCKSGFYLHL GKCLDNCPEG LEANNHTMEC VSIVHCEVSE WNPWSPCTKK GKTCGFKRGT ETRVREIIQH PSAKGNLCPP TNETRKCTVQ RKKCQKGERG KKGRERKRKK PNKGESKEAI PDSKSLESSK EIPEQRENKQ QQKKRKVQDK QKSVSVSTVH
Content And Storage -20°C
Activity R-Spondin-3 enhances BMP-2-mediated differentiation of MC3T3-E1 cells. The expected ED50 for this effect is 0.8 -2.0 ug/ml.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.