Learn More
ACROBiosystems ACROBiosystems Human R-Spondin 3 / RSPO3 Protein, Fc Tag
Catalog No. 502107346
Encompass_Preferred
Recombinant Protein;1MG;Human R-Spondin 3 Protein, Fc Tag (RS3-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 272 (Accession # Q9BXY4-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 55 kDa.
Catalog No. 50-210-7346 Supplier ACROBiosystems Supplier No. RS3H52531MG
Recombinant Protein;1MG;Human R-Spondin 3 Protein, Fc Tag (RS3-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 272 (Accession # Q9BXY4-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 55 kDa.