Learn More
Biomatik Corporation Human RAB1A, Member RAS Oncogene Family (RAB1A) CLIA Kit
Supplier:  Biomatik Corporation EKU10016
Encompass_Preferred
Human RAB1A, Member RAS Oncogene Family (RAB1A) CLIA Kit for detecting RAB1A in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P62820. Gene ID: 5861. Target Synonyms: RAB1; YPT1; Ras-related protein Rab-1A; YPT1-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5377