Learn More
Sino Biological Anti-RAB27B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 16007T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RAB27B (rh RAB27B; Catalog#16007-H07H; NP_004154.2; Thr2-Lys215). RAB27B specific IgG was purified by Human RAB27B affinity chromatography.
Catalog No. 50-255-4075