Learn More
Biomatik Corporation Human RAB37, Member RAS Oncogene Family (RAB37) ELISA Kit
Supplier:  Biomatik Corporation EKU07001
Encompass_Preferred
Human RAB37, Member RAS Oncogene Family (RAB37) ELISA Kit for detecting RAB37 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96AX2. Gene ID: 326624. Target Synonyms: Ras-related protein Rab-37. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8790