Learn More
Biomatik Corporation Human RAB5A, Member RAS Oncogene Family (RAB5A) CLIA Kit
Supplier:  Biomatik Corporation EKU09908
Encompass_Preferred
Human RAB5A, Member RAS Oncogene Family (RAB5A) CLIA Kit for detecting RAB5A in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.05-38ng/ml. Sensitivity: 0.02ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P20339. Gene ID: 5868. Target Synonyms: RAB5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6150