Learn More
Sino Biological Anti-C20orf24 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103623T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human C20orf24 / RAB5IF, and purified by antigen affinity chromatography.
Catalog No. 50-240-7736