Learn More
Biomatik Corporation Human Rac-GTPase Activating Protein 1 (RACGAP1) ELISA Kit
Supplier:  Biomatik Corporation EKU07005
Encompass_Preferred
Human Rac-GTPase Activating Protein 1 (RACGAP1) ELISA Kit for detecting RACGAP1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H0H5. Gene ID: 29127. Target Synonyms: HsCYK-4; ID-GAP; MgcRacGAP; Male germ cell RacGap. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7122