Learn More
Sino Biological Anti-Rack1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12498T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RACK1/GNB2L1 (rh RACK1/GNB2L1; Catalog#12498-H10E; P63244; Met1-Arg317). RACK1/GNB2L1 specific IgG was purified by Human RACK1/GNB2L1 affinity chromatography.
Catalog No. 50-255-4368