Learn More
Sino Biological Anti-ULBP4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 16073RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RAET1E / ULBP4 (rh RAET1E / ULBP4; Catalog#16073-H08H; NP_631904.1; Met1-Asp225). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4061