Learn More
Sino Biological Anti-ULBP6/RAET1L Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 15759RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RAET1L (rh RAET1L; Catalog#15759-H08H; AAK91503.1; Met1-Ser217). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4085