Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RAI2 Partial ORF (NP_068557.1, 431 a.a. - 530 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959299 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-299 10 μg
89-959-300 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-299 Supplier Abnova™ Supplier No. H00010742Q01S

missing translation for 'validMainframeMsg'

Used for AP, Array, ELISA, WB-Re

Retinoic acid plays a critical role in development, cellular growth, and differentiation. The specific function of this retinoic acid-induced gene has not yet been determined; however, it has been suggested to play a role in development. Localization of this gene designates it to be a candidate for diseases such as Nance-Horan syndrome, sensorineural deafness, non-specific X-linked mental retardation, oral-facial-digital syndrome, and Fried syndrome. [provided by RefSeq]

Sequence: VGESQAAKVIVSVEDAVPTIFCGKIKGLSGVSTKNFSFKREDSVLQGYDINSQGEESMGNAEPLRKPIKNRSIKLKKVNSQEIHMLPIKKQRLATFFPRK

Specifications

Accession Number NP_068557.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10742
Molecular Weight (g/mol) 36.74kDa
Name RAI2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen VGESQAAKVIVSVEDAVPTIFCGKIKGLSGVSTKNFSFKREDSVLQGYDINSQGEESMGNAEPLRKPIKNRSIKLKKVNSQEIHMLPIKKQRLATFFPRK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name RAI2
Gene Symbol RAI2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.