Learn More
Biomatik Corporation Human RalA Binding Protein 1 (RALBP1) ELISA Kit
Supplier:  Biomatik Corporation EKU07009
Encompass_Preferred
Human RalA Binding Protein 1 (RALBP1) ELISA Kit for detecting RALBP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15311. Gene ID: 10928. Target Synonyms: RIP; RIP1; RLIP76; 76 kDa Ral-interacting protein; Ral-interacting protein 1; Dinitrophenyl S-glutathione ATPase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7163