Learn More
Biomatik Corporation Human RALBP1 Associated Eps Domain Containing Protein 1 (REPS1) ELISA Kit
Supplier:  Biomatik Corporation EKU12103
Encompass_Preferred
Human RALBP1 Associated Eps Domain Containing Protein 1 (REPS1) ELISA Kit for detecting REPS1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96D71. Gene ID: 85021. Target Synonyms: RalBP Associated Eps15-Homology Domain protein; RalBP1-interacting protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8027