Learn More
FUJIFILM Biosciences Human RANK-Ligand, Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Catalog No. 50990713
Encompass_Preferred
Receptor activator of nuclear factor kappa-B Ligand (RANK Ligand) is a cell-bound marker related to the tumor necrosis factor (TNF) family of proteins. RANK Ligand plays a critical role in bone metabolism and osteoclast differentiation. T cell expression of RANK Ligand promotes dendritic cell maturation.
Catalog No. 50-990-713 Supplier FUJIFILM Biosciences Supplier No. 1001910UG
Add to Cart
Edge
Add to Cart
Receptor activator of nuclear factor kappa-B Ligand (RANK Ligand) is a cell-bound marker related to the tumor necrosis factor (TNF) family of proteins. RANK Ligand plays a critical role in bone metabolism and osteoclast differentiation. T cell expression of RANK Ligand promotes dendritic cell maturation.