Learn More
Sino Biological Human RANKL/TNFSF11 Matched ELISA Antibody Pair Set 5 Plates
Supplier: Sino Biological SEK116825
The Human RANKL / OPGL / TNFSF11 / CD254 ELISA Pair Set is for the quantitative determination of Human RANKL / OPGL / TNFSF11 / CD254.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human RANKL / OPGL / TNFSF11 / CD254 coated on a 96-well plate. Standards and samples are added to the wells, and any Human RANKL / OPGL / TNFSF11 / CD254 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human RANKL / OPGL / TNFSF11 / CD254 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human RANKL / OPGL / TNFSF11 / CD254 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.