Learn More
Biomatik Corporation Human RAR Related Orphan Receptor Gamma (RORg) ELISA Kit
Supplier:  Biomatik Corporation EKU07010
Encompass_Preferred
Human RAR Related Orphan Receptor Gamma (RORg) ELISA Kit for detecting RORg in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51449. Gene ID: 6097. Target Synonyms: RORC; NR1F3; RORG; RZRG; TOR; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan receptor C; Retinoid-related orphan receptor-gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8787