Learn More
Bioworld HUMAN RAS HOMOLOG ENRICHED
Supplier:  Bioworld 220605021
Encompass_Preferred
HUMAN RAS HOMOLOG ENRICHED IN BRAIN (RECOMBINANT): Recombinant Human TransGlutaminase II Source: E.coli Presentation: 100 l purified human Transglutaminase II protein at 100 g/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol...
Catalog No. 50-253-597