Learn More
Biomatik Corporation Human Ras Related C3 Botulinum Toxin Substrate 1 (Rac1) CLIA Kit
Supplier:  Biomatik Corporation EKU12257
Encompass_Preferred
Human Ras Related C3 Botulinum Toxin Substrate 1 (Rac1) CLIA Kit for detecting Rac1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P63000. Gene ID: 5879. Target Synonyms: TC-25; p21-Rac1; MIG5; Cell migration-inducing gene 5 protein; Ras-like protein TC25; p21-Rac1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5124