Learn More
Sino Biological Anti-RbAp48 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11309T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RbAp48 (rh RbAp48; Catalog#11309-H07B; NP_005601.1; Met1-Ser425). RbAp48 specific IgG was purified by Human RbAp48 affinity chromatography.
Catalog No. 50-255-4713