Learn More
AURORA BIOLABS LLC Human RBD-RAF1, 100 μg
Catalog No. NC3787944
Encompass
Human Ras Binding Domain (RBD) of Raf-1 Proto-Oncogene, Serine/Threonin Kinase (RAF1), (UniProt # P04049, amino acids 50-140), expressed with an N-terminal 6XHis and C-terminal FLAG in E. coli. Specifically binds to and precipitates Ras-GTP from cell lysate. Can be bound to glutathione coated plates prior to precipitating Ras-GTP.
Catalog No. NC3787944 Supplier AURORA BIOLABS LLC Supplier No. 7237231100
Human Ras Binding Domain (RBD) of Raf-1 Proto-Oncogene, Serine/Threonin Kinase (RAF1), (UniProt # P04049, amino acids 50-140), expressed with an N-terminal 6XHis and C-terminal FLAG in E. coli. Specifically binds to and precipitates Ras-GTP from cell lysate. Can be bound to glutathione coated plates prior to precipitating Ras-GTP.