Learn More
Raybiotech Inc Human RBP4 ELISA Kit
Supplier:  Raybiotech Inc ELHRBP41
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human RBP4. This antibody pair does not cross-react with mouse RBP4.
Catalog No. 50-194-9677