Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RCAN2 (aa 4-30) Control Fragment Recombinant Protein

Catalog No. RP101503
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101503 Supplier Invitrogen™ Supplier No. RP101503
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62860. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Regulator of calcineurin 2 (RCAN2), also known as ZAKI4 and DSCR1L1, is expressed as two isoforms differing at their N-terminus. The longer of the two (isoform 1) is expressed exclusively in the brain, while isoform 2 is ubiquitously expressed, with highest expression in brain, heart, and muscle. Both isoforms bind to the catalytic subunit of calcineurin, a Ca++-dependent protein phosphatase involved in several neuronal functions, though their C-terminal region and inhibit calcineurin's activity. Unlike isoform 1 of RCAN2, the expression of the second isoform is not induced by the thyroid hormone T3. RCAN2 is a member of a family of three endogenous calcineurin regulators that are located near the minimal supernumerary fragment of chromosome 21 in individuals with Down syndrome, suggesting that they play a role in this syndrome. Multiple isoforms of RCAN2 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14206
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 10231
Name Human RCAN2 (aa 4-30) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias calcineurin inhibitory protein ZAKI-4; Calcipressin-2; caskin-1-like; CSP2; Dcip2; Down syndrome candidate region 1-like 1; down syndrome candidate region 1-like protein 1; Down syndrome critical region gene 1-like 1; Down syndrome critical region gene 1-like 1 protein; Down syndrome critical region gene1-like 1; DSCR1L1; DSCR1L1 protein; LOW QUALITY PROTEIN: calcipressin-2; MCIP2; myocyte-enriched calcineurin interactin protein 2; myocyte-enriched calcineurin-interacting protein 2; Rcan2; RCN2; regulator of calcineurin 2; si:ch211-124g24.1; thyroid hormone-responsive (skin fibroblasts); thyroid hormone-responsive protein ZAKI-4; translation initiation factor IF-2; ZAKI4; ZAKI-4; zgc:85746
Common Name RCAN2
Gene Symbol Rcan2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PSMDCDVSTLVACVVDVEVFTNQEVKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.