Learn More
Biomatik Corporation Human RCC1 And BTB Domain Containing Protein 2 (RCBTB2) ELISA Kit
Supplier:  Biomatik Corporation EKU07015
Encompass_Preferred
Human RCC1 And BTB Domain Containing Protein 2 (RCBTB2) ELISA Kit for detecting RCBTB2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95199. Gene ID: 1102. Target Synonyms: CHC1L; RLG; RCC1-like G exchanging factor; Regulator Of Chromosome Condensation And BTB (POZ) Domain Containing Protein 2; Chromosome Condensation 1-Like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8786