Learn More
Cell Signaling Technology Human Reactive DNA Demethylation Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502730748
Encompass
The Human Reactive DNA Demethylation Antibody Sampler Kit offers an economical solution for the detection of TET protein family members and TDG protein. This kit is designed to provide sufficient antibody quantity for performing two western blot experiments with each primary antibody included, facilitating the study of active cytosine demethylation and related epigenetic pathways.
  • Includes primary antibodies for TET1, TET2, TET3, and TDG
  • Supplied with HRP-linked anti-rabbit IgG secondary antibody
  • Validated for multiple applications including western blotting, immunoprecipitation, and chromatin analysis
  • Economical format for assessing DNA demethylation regulatory proteins
Catalog No. 50-273-0748 Supplier Cell Signaling Technology Supplier No. 84884T
The Human Reactive DNA Demethylation Antibody Sampler Kit offers an economical solution for the detection of TET protein family members and TDG protein. This kit is designed to provide sufficient antibody quantity for performing two western blot experiments with each primary antibody included, facilitating the study of active cytosine demethylation and related epigenetic pathways.
  • Includes primary antibodies for TET1, TET2, TET3, and TDG
  • Supplied with HRP-linked anti-rabbit IgG secondary antibody
  • Validated for multiple applications including western blotting, immunoprecipitation, and chromatin analysis
  • Economical format for assessing DNA demethylation regulatory proteins