Learn More
Cell Signaling Technology Human Reactive PANoptosis Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502727012
Encompass
The Human Reactive PANoptosis Antibody Sampler Kit offers a cost-effective solution for detecting PANoptosis pathway activation in human samples. This comprehensive kit contains a selection of primary antibodies designed to identify key proteins and their phosphorylated or cleaved states associated with pyroptosis, apoptosis, and necroptosis. Each antibody is provided in a quantity sufficient for two western blot experiments.
  • Includes antibodies for detection of IL-1β, RIPK3, gasdermin D, MLKL, and Caspase-3, including specific phosphorylated and cleaved forms.
  • Supplied with HRP-linked anti-rabbit IgG secondary antibody.
  • Validated for multiple applications including western blotting, immunofluorescence, immunohistochemistry (paraffin), flow cytometry, and immunoprecipitation.
  • Storage at −20°C required; antibodies should not be aliquoted.
Catalog No. 50-272-7012 Supplier Cell Signaling Technology Supplier No. 34886T
The Human Reactive PANoptosis Antibody Sampler Kit offers a cost-effective solution for detecting PANoptosis pathway activation in human samples. This comprehensive kit contains a selection of primary antibodies designed to identify key proteins and their phosphorylated or cleaved states associated with pyroptosis, apoptosis, and necroptosis. Each antibody is provided in a quantity sufficient for two western blot experiments.
  • Includes antibodies for detection of IL-1β, RIPK3, gasdermin D, MLKL, and Caspase-3, including specific phosphorylated and cleaved forms.
  • Supplied with HRP-linked anti-rabbit IgG secondary antibody.
  • Validated for multiple applications including western blotting, immunofluorescence, immunohistochemistry (paraffin), flow cytometry, and immunoprecipitation.
  • Storage at −20°C required; antibodies should not be aliquoted.