Learn More
Raybiotech Inc G-Series Human Receptor Array 1 Kit.
Supplier:  Raybiotech Inc GSHREC12
Encompass_Preferred
Semi-quantitative measurement of 40 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: 4-1BB (TNFRSF9/CD137), ALCAM (CD166), BCMA (TNFRSF17), CD14, CD30 (TNFRSF8), CD40 Ligand (TNFSF5), CD80 (B7-1), CEACAM-1, DR6 (TNFRSF21), Dtk, Endoglin (CD105), ErbB3, E-Selectin, Fas (TNFRSF6/Apo-1), Flt-3 Ligand, GITR (TNFRSF18), HVEM (TNFRSF14), ICAM-3 (CD50), IL-1 R1, IL-1 R4 (ST2), IL-10 R beta, IL-17 RA, IL-2 R gamma (Common gamma Chain), IL-21 R, LIMPII, Lipocalin-2 (NGAL), L-Selectin (CD62L), LYVE-1, MICA, MICB, NRG1-beta 1 (HRG1-beta 1), PDGF R beta, PECAM-1 (CD31), RAGE, TIM-1 (KIM-1), TRAIL R3 (TNFRSF10C), Trappin-2, uPAR, VCAM-1 (CD106), XEDAR
Catalog No. 50-195-0419