Learn More
Apexbio Technology LLC GM-CSF, human recombinant(Synonyms: Granulocyte-Macrophage Colony-Stimulating Factor, GM-CSF, Recombinant Human GM-CSF, rhGM-CSF), 100ug.
SDP
Catalog No. 5000061133
Encompass_Preferred
Human recombinant granulocyte-macrophage colony-stimulating factor (GM-CSF) is a cytokine that stimulates the proliferation and differentiation of myeloid progenitor cells into granulocytes and macrophages. Secreted by activated immune cells such as T cells, macrophages, endothelial cells, and fibroblasts, GM-CSF promotes hematopoietic cell colony formation in vitro. Additionally, GM-CSF has been shown to regulate migration and proliferation in non-hematopoietic cells, including endothelial and certain tumor cell lines. Clinically, recombinant human GM-CSF is utilized to stimulate leukocyte recovery after chemotherapy and explored as an immunological adjuvant in vaccine trials for HIV-infected patients.
Catalog No. 50-000-61133 Supplier Apexbio Technology LLC Supplier No. P10120.1
Human recombinant granulocyte-macrophage colony-stimulating factor (GM-CSF) is a cytokine that stimulates the proliferation and differentiation of myeloid progenitor cells into granulocytes and macrophages. Secreted by activated immune cells such as T cells, macrophages, endothelial cells, and fibroblasts, GM-CSF promotes hematopoietic cell colony formation in vitro. Additionally, GM-CSF has been shown to regulate migration and proliferation in non-hematopoietic cells, including endothelial and certain tumor cell lines. Clinically, recombinant human GM-CSF is utilized to stimulate leukocyte recovery after chemotherapy and explored as an immunological adjuvant in vaccine trials for HIV-infected patients.