Learn More
STEMCELL Technologies Human Recombinant EGF, ACF, Size: 1000 µg
SDP
Supplier:  STEMCELL Technologies 78136.2
Encompass
Epidermal growth factor (EGF) is characterized by high-affinity binding to various EGF receptors (EGFRs) and the production of mitogenic responses (Carpenter & Cohen). EGF promotes EGFR dimerization, resulting in activation of downstream pathways including PI3K, ERK1/2, JAK/STAT, β-catenin, and calcium signaling. EGF is secreted by the gut-associated salivary and Brunner’s glands, is found in a variety of body fluids, and stimulates cell proliferation and differentiation in rodent and neonatal human intestine (Wright et al.). Central nervous system stem cells also proliferate in response to the EGF stimulus (Reynolds & Weiss). This product is animal component-free.
Catalog No. 50-197-6554