Learn More
STEMCELL Technologies Human Recombinant EPO, Size: 10 µg
SDP
Supplier:  STEMCELL Technologies 78007.1
Encompass
Erythropoietin (EPO) is a glycoprotein growth factor that is produced primarily in the kidney in response to hypoxia or anemia. It is the principal physiological regulator of erythropoiesis. EPO promotes erythropoiesis by binding to a homodimeric cell surface receptor that activates JAK2/STAT5, PI3K/AKT, and MAPK pathways, and stimulates the proliferation and differentiation of erythroid progenitor cells (Jelkmann; Kuhrt & Wojchowski).
Catalog No. 50-197-6256
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov