Learn More
STEMCELL Technologies Human Recombinant FGF-7, ACF, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 78186.1
Encompass
Fibroblast growth factor 7 (FGF-7) is a member of the FGF family, and acts exclusively through a subset of FGF receptor isoforms expressed predominantly by epithelial cells (Finch & Rubin). FGF-7 seems to act specifically on epithelial cells and stimulates proliferation, migration, and differentiation of these cells, and also participates in epithelial protection and repair both in vitro and in vivo (Finch & Rubin; Werner). In contrast, FGF-7 is produced solely by cells of mesenchymal origin, and functions as a paracrine mediator of mesenchymal-epithelial communication (Rubin et al.). FGF-7 has also been shown to supplement several wound-healing properties of bioengineered skin (Erdag et al.) and to induce autophagy in human keratinocytes (Belleudi et al.). Additionally, FGF-7 has a role in the differentiation of pluripotent stem cell to endodermal pancreatic-like insulin-producing cells and thymic epithelial cells (Inami et al.; Niu et al.). This product is animal component-free.
Catalog No. 50-197-6676